Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kelch-Like 3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303236100UL
Description
Kelch-Like 3 Polyclonal specifically detects Kelch-Like 3 in Human, Mouse samples. It is validated for Western Blot.Specifications
Kelch-Like 3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
26249 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
FLJ40871, kelch-like 3 (Drosophila), kelch-like protein 3, KIAA1129kelch (Drosophila)-like 3, MGC44594 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human Kelch-Like 3 (NP_059111.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction