Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kelch Like Family Member 42 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259107
Description
KLHDC5 Polyclonal specifically detects KLHDC5 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Kelch Like Family Member 42 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
KLHL42 | |
Maps to a region between 400 and 450 | |
100 μL | |
Primary | |
Store at 4C. |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction