Learn More
Sklar InstrumentsSupplier Diversity Partner SklarTech 5000 Kelly Forceps, Sklar, Blunt/Blunt, Curved, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 319043
SklarTech 5000 Kelly Forceps, Sklar, Blunt/Blunt, Curved, Serrated, Stainless Steel. Sklartech 5000 Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue during laparoscopic procedures. They may also be used for soft tissue dissection. They are available curved or straight with fully serrated jaws. Sklartech 5000 Laparoscopic instruments are crafted from the highest grade stainless steel to ensure reliability and durability. Sklar offers laparoscopic instruments in the most widely used patterns, as complete sets or replacement parts only. Each complete set includes a handle, locking collar, insulated shaft, and insert with the tip. Sklartech 5000 replacement parts are interchangeable based on procedure or surgeon preference. Miniature replacement parts are also available. This product is a replacement part that is straight with 5mm serrated tips and a length of 33cm.
Catalog No. 50-143-8164