Learn More
Sklar InstrumentsSupplier Diversity Partner Sklarlite Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Blunt, Curved, Serrated, Extra Light, Stainless Steel. Sklarlite Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 232108
Sklarlite Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Blunt, Curved, Serrated, Extra Light, Stainless Steel. Sklarlite Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Sklarlite instruments are a lighter version of standard Sklar instruments. This lightweight modification provides the surgeon with greater control and less fatigue. This product is curved with serrated tips and a length of 5-1/2 inches.
Catalog No. 50-143-7956