Learn More
Sklar InstrumentsSupplier Diversity Partner Sklarlite XD Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Curved, Serrated, Extra Delicate, EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 232508
Sklarlite XD Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Curved, Serrated, Extra Delicate, Extra Delicate. Sklarlite XD Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Sklarlite XD instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. This product is curved with serrated tips and a length of 5-1/2 inches.
Catalog No. 50-143-7981