Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Kelly-Rankin Hemostatic Forceps, Sklar, 6-1/4 in., Blunt, Curved, Serrated, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21469
Econo Kelly-Rankin Hemostatic Forceps, Sklar, 6-1/4 in., Blunt, Curved, Serrated, Stainless Steel. Econo Kelly-Rankin Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. The jaws are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Standard Kelly forceps are available in two lengths, 5-1/2 inches and 7 inches. Kelly forceps that have an overall length of 6-1/4 inches are referred to as Kelly-Rankin forceps. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with serrated tips and a length of 5-1/2 inches. They are sold in boxes of 12.
Catalog No. 50-143-7639