Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Build a Crystal Radio w/an Amplifier: Exploring Wavelength, Frequency, Amplitude
SCI_ED

Students explore wavelength, frequency and amplitude while immersed in hands-on experimentation.

$545.00

Specifications

For Use With (Application) Ideal for hands on learners, as intuition plays a role in developing design solutions, iterative testing and making modifications for improvements
Product Type Physics Classroom Kit
Min. Grade Level 7
Max. Grade Level 10
View More Specs

Alert:

Choking hazard (1) Small parts. Not for children under 3 yrs.


Includes: Complete instructor's manual with reproducible stepwise student protocols.

Requires: Impact goggles


Products 1
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
S07355
SCI_ED
View Documents Promotion Details
Aqua Phoenix Scientific
15230
Complete instructor's manual with reproducible stepwise student protocols, enough materials to build six radios with amplifiers
Each for $545.00
Add to cart
 
Description

Description

Students build their own old fashioned Crystal Radios and explore the effects of design variations within the coils, building materials, and antennae number and length, on maximizing tuning and signal strength.

  • Students upgrade their crystal radio into transistor radio
  • Ideal for hands on learners, as intuition plays a role in developing antenna design solutions, iterative testing, and making modifications for improvements (iterative cycle)
  • Great for introducing guided inquiry-based learning
  • Grade 7 to 10
  • Available as single kit (1 to 4 students) or class pack kit (24 students)

Order Info

Single-Pack includes enough materials to build one radio with an amplifier.
Class-Pack includes enough materials to build six radios with amplifiers.

Specifications

Specifications

Ideal for hands on learners, as intuition plays a role in developing design solutions, iterative testing and making modifications for improvements
Physics Classroom Kit
7
10
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit