Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Build a Crystal Radio w/an Amplifier: Exploring Wavelength, Frequency, Amplitude Products for Science Education

Students explore wavelength, frequency and amplitude while immersed in hands-on experimentation.

Manufacturer:  Aqua Phoenix Scientific 15230

 View more versions of this product

Catalog No. S07355


Add to cart

Alert:

Choking hazard (1) Small parts. Not for children under 3 yrs.


Includes: Complete instructor's manual with reproducible stepwise student protocols.

Requires : Impact goggles

Description

Description

Students build their own old fashioned Crystal Radios and explore the effects of design variations within the coils, building materials, and antennae number and length, on maximizing tuning and signal strength.

  • Students upgrade their crystal radio into transistor radio
  • Ideal for hands on learners, as intuition plays a role in developing antenna design solutions, iterative testing, and making modifications for improvements (iterative cycle)
  • Great for introducing guided inquiry-based learning
  • Grade 7 to 10
  • Available as single kit (1 to 4 students) or class pack kit (24 students)

Order Info

Single-Pack includes enough materials to build one radio with an amplifier.
Class-Pack includes enough materials to build six radios with amplifiers.

Specifications

Specifications

Ideal for hands on learners, as intuition plays a role in developing design solutions, iterative testing and making modifications for improvements
Physics Classroom Kit
7
Complete instructor's manual with reproducible stepwise student protocols, enough materials to build six radios with amplifiers
10
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit