Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Build Your Own Simple Electric Motor Kit
SCI_ED

Build your own simple electric motors using wire and magnets.

$69.75 - $239.00

Specifications

For Use With (Application) Designed for whole class to extend learning from electromagnets to motors
Product Type Physics Classroom Kit
Min. Grade Level 5
Max. Grade Level 8
View More Specs

Alert:

Choking hazard (1) Small parts. Not for children under 3 yrs.


Includes: Magnet wire, copper wire, rectangular magnets, D-Cell battery holder w/leads, round magnets, small nails, ring magnet, wire cutter, neodymium magnet, sandpaper, dowel rods, instructor's manual

Requires: Safety goggles and batteries (three D-cell for single-pack kit and 18 D-cell for class-pack kit).


Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
S07350
SCI_ED
View Documents
Aqua Phoenix Scientific
15212
4 experiments, background information, sample data and complete answer keys, instructor's manual, 6 motors
Each for $239.00
Add to cart
 
S07349
SCI_ED
View Documents
Aqua Phoenix Scientific
15210
4 experiments, background information, sample data and complete answer keys, instructor's manual
Each for $69.75
Add to cart
 
Description

Description

  • Hands-on project set designed for whole class to extend learning from electromagnets to motors
  • Great for introducing guided inquiry-based learning, systematic problem solving and teaching engineering design practices
  • Explore effects of altering coils in motor, wire thickness and effects of using different sizes, shapes, strengths and types of magnets
  • Includes instructor's manual with reproducible stepwise student protocols
  • Includes four experiments which culminate in a design activity
  • With instructor's manuals with lesson plans, background information, reproducible stepwise student protocols and guided worksheets
  • Availalbe as single kit (1 to 4 students) or class pack kit (24 students)
  • Grade 5 to 8

Specifications

Specifications

Designed for whole class to extend learning from electromagnets to motors
Physics Classroom Kit
5
8
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit