Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Let's Build Electromagnets Kit
SCI_ED

Introduce the Engineering Design Process for problem solving as you explore electricity and magnetism in this electromagnet building exercise.

$62.50 - $128.00

Specifications

For Use With (Application) Introduce students to electricity and magnetism with this electromagnet building exercise
Grade 3 to 5
Includes Instructor's manual, student guide, worksheets, batteries and all materials required for investigation
View More Specs

Alert:

Choking hazard (1) Small parts. Not for children under 3 yrs.


Includes: Dowel rod, alligator clip leads, 6V lantern battery, wire strippers, magnet wire, masking tape, paper clips, steel washers, steel nails, sandpaper, steel bolts, weigh dish, instructor's manual, reproducible student data sheets

Requires: Impact goggles


Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S07348
SCI_ED
View Documents
Aqua Phoenix Scientific
15205
Class pack kit
Each for $128.00
Add to cart
 
S07347
SCI_ED
View Documents
Aqua Phoenix Scientific
15201
Single kit
Each for $62.50
Add to cart
 
Description

Description

  • Explore magnetic field, electrical charge, and effects of increasing current on the generation of heat
  • Compare core materials, core thickness, and number of wraps in coils in inquiry based investigations
  • Four engaging activities culminate with students applying what they learn in a design the strongest electromagnet activity
  • Instructor's manual with lesson plans, background information, reproducible stepwise student instructions, worksheets, and complete answer keys
  • Grades 3 to 5
  • Available as single kit (1 to 4 students) or class pack kit (24 students)

Specifications

Specifications

Introduce students to electricity and magnetism with this electromagnet building exercise
3 to 5
Instructor's manual, student guide, worksheets, batteries and all materials required for investigation
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit