Learn More
Ken-A-Vision™ Multicharger 3 Microscope Power Port Products for Science Education
Port keeps microscopes fully-charged and ready for classroom projects and assignments
Manufacturer:  Ken-A-Vision, Inc. SCGN061U
Catalog No. S21061
  Check Availability
Add to cart
Specifications
Description
  • Allows one electrical wall outlet to charge eight microscopes at once
  • LED meter indicators show percentage of charge received
  • Red and green LED station indicators show when microscope is charging or fully charged
  • Provides means to charge multiple microscopes while they are in storage
Description & Specifications
Specifications
8 Plugs
4 lbs.
14 x 21.6 x 4.5cm (5.5 x 8.5 x 1.8 in.)
Ken-A-Vision Microscopes
1.8kg