Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratan Sulfate Mouse anti-Human, Clone: 5-D-4, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen Keratan Sulfate
Clone 5-D-4
Concentration 0.5 mg/mL
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN2483100
View Documents
MilliporeSigma
MABN2483100UL
100 μL
Each for $373.00
Add to cart
 
MABN248325
View Documents
MilliporeSigma
MABN248325UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

Keratan Sulfate Monoclonal antibody specifically detects Keratan Sulfate in Human samples. It is validated for ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blotting.
Specifications

Specifications

Keratan Sulfate
0.5 mg/mL
Unconjugated
Mouse
Human
Proteoglycan core protein isolated after chondroitinase ABC digestion of human articular cartilage proteoglycan monomer.
Monoclonal
Protein G purified
5-D-4
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit