Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin, Powder, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
69430-36-0
C2H2BrClO2
173.39
GEHJBWKLJVFKPS-UHFFFAOYNA-N
2-bromo-2-chloroacetic acid
OC(=O)C(Cl)Br
69430-36-0
C2H2BrClO2
173.39
GEHJBWKLJVFKPS-UHFFFAOYNA-N
2-bromo-2-chloroacetic acid
OC(=O)C(Cl)Br

K3030

Spectrum™ Keratin, Powder is the key material that makes up the human body’s out layer of skin. It is also the key element of hair and nails. It is considered to be similar to chitin. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Poly Bottle
Quantity 100 g
CAS Max % 1