Learn More
Keratin31 Mouse anti-Human, Janelia Fluor 646, Clone: LHTric-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP253114JF646
Catalog No. NBP53114646
Add to cart
Description
Description
Keratin31 Monoclonal antibody specifically detects Keratin31 in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
Keratin31
LHTric-1
Immunohistochemistry
Purified
KRT31
CVPRPRCGPCNSFVR
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human, Rat
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Hair Keratin, Type I Ha, Hard Keratin, Type I, 1, hHa1, K31, Keratin 31, Keratin, Hair, Acidic, 1, Keratin, Type I Cuticular Ha1, Keratin-31, KRTHA1
Mouse
IgG2a
0.1mL
Primary
3881