Learn More
Gibco™ Keratinocyte AOF Growth Kit Non-distribution product as customer accommodation.
Keratinocyte AOF Growth Kit
Manufacturer:  Gibco™ A1051501
Catalog No. A1051501
Add to cart
Description
Description
Growth kit for the animal origin-free culture of primary human keratinocytes. Contains 1 each of EpiLife Basal Medium (M-EPI-500-CA), Supplement S7 (S-017-5), and Coating Matrix Kit (R-011-K).
Specifications
Specifications
Keratinocytes
Mammalian cell culture, primary cell culture, Keratinocyte Culture, cell culture
1 Kit(s)
1 Kit(s)
Keratinocyte AOF Growth Kit, 1 Kit, Keratinocytes Cell Type, Human Species, Suspension Cell Culture Type, Animal Origin-Free, Chemically Defined, Serum-Free Classification, For Mammalian Cell Culture, Primary Cell Culture
1 Each of epiLife basal medium (M-EPI-500-CA), supplement S7 (S-017-5) and coating matrix kit (R-011-K). Store each component at the temperature indicated on the label. Store each component at the temperature indicated on the label
Keratinocyte AOF Growth Kit
For research use only
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.