Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketanserin tartrate, Tocris Bioscience™

Click to view available options
Quantity:
50mg
250mg

Chemical Identifiers

83846-83-7
C26H28FN3O9
545.52
KMTLTEVOQLMYRS-WUUYCOTASA-N
3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2, 4[1H, 3H]-quinazolinedione tartrate
91885469

Selective 5-HT2A antagonist. Also antagonist at 5-HT1D

Selective 5-HT2A serotonin receptor antagonist; can also be used to discriminate between 5-HT1D and 5-HT1B receptor subtypes.
SMILES C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O.C(C(C(=O)O)O)(C(=O)O)O
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid;3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
Formula Weight 545.52
Quantity 250mg
Chemical Name or Material Ketanserin tartrate
Grade >97%

Recommended Storage : Store at Room Temperature