Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoconazole, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog Number 18603641
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
65277-42-1
C26H28Cl2N4O4
531.43
XMAYWYJOQHXEEK-ZEQKJWHPSA-N
1-[4-(4-{[(2S,4R)-2-(2,4-dichlorophenyl)-2-[(1H-imidazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]ethan-1-one
CC(=O)N1CCN(CC1)C1=CC=C(OC[C@@H]2CO[C@](CN3C=CN=C3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1
65277-42-1
C26H28Cl2N4O4
531.43
XMAYWYJOQHXEEK-ZEQKJWHPSA-N
1-[4-(4-{[(2S,4R)-2-(2,4-dichlorophenyl)-2-[(1H-imidazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]ethan-1-one
CC(=O)N1CCN(CC1)C1=CC=C(OC[C@@H]2CO[C@](CN3C=CN=C3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1

K1149, 65277-42-1, C26H28Cl2N4O4

Spectrum™ Chemical Ketoconazole, USP is a synthetic antifungal medication that is used primarily to treat fungal infections. While its use as an oral tablet has been discontinued in many countries due to concerns that it caused serious liver injury, it is still used in a variety of topical administrations such as creams, which are used to treat tinea, candidal paronychia, and pityriasis versicolor and in shampoos used primarily to treat seborrheic dermatitis (dandruff). The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Poly Pail
Quantity 2.5 kg
UN Number UN2811
Specific Rotation -1° to +1°
Percent Purity 98 to 102%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit