Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketohexokinase Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI1D8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271069APCCY7

Catalog No. N271069APC7


Add to cart

Description

Description

Ketohexokinase Monoclonal antibody specifically detects Ketohexokinase in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Ketohexokinase
OTI1D8
Flow Cytometry
Affinity Purified, Conjugated
KHK
Full length recombinant protein of human Ketohexokinase (NP_000212) produced in HEK293T cells.
Immunogen affinity purified
Lipid and Metabolism
Primary
3795.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
EC 2.7.1.3, Hepatic fructokinase, ketohexokinase, ketohexokinase (fructokinase)
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit