Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoprofen, USP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
22071-15-4
C16H14O3
254.29
DKYWVDODHFEZIM-UHFFFAOYNA-N
2-(3-benzoylphenyl)propanoic acid
CC(C(O)=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
22071-15-4
C16H14O3
254.29
DKYWVDODHFEZIM-UHFFFAOYNA-N
2-(3-benzoylphenyl)propanoic acid
CC(C(O)=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1

K1155, 22071-15-4, C16H14O3

Spectrum™ Ketoprofen, USP is a nonsteroidal anti-inflammatory drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Ignition Residue 0.002
Loss on Drying 0.005
Specific Rotation +1 to -1°
Quantity 1 kg
Melting Range 92.0-97.0°C
CAS Max % 1
UN Number UN2811
Percent Purity 98.5 to 101%