Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ketoprofen, USP, 98.5-101%, Spectrum™ Chemical

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Description
Spectrum™ Chemical Ketoprofen, USP is a nonsteroidal anti-inflammatory drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Ignition Residue | 0.2% |
Loss on Drying | 0.5% |
Melting Range | 92.0-97.0°C |
UN Number | UN2811 |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Specific Rotation | +1° to -1° |
Percent Purity | 98.5 to 101% |
Grade | USP |