Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketorolac Tromethamine, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
74103-07-4
C19H24N2O6
376.41
BWHLPLXXIDYSNW-UHFFFAOYNA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1
74103-07-4
C19H24N2O6
376.41
BWHLPLXXIDYSNW-UHFFFAOYNA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol; 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
NC(CO)(CO)CO.OC(=O)C1CCN2C1=CC=C2C(=O)C1=CC=CC=C1

K1204, 74103-07-4, C15H13NO3•C4H11NO3

Spectrum™ Ketorolac Tromethamine, USP is a nonsteroidal anti-inflammatory drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Quantity 100 g
Loss on Drying 0.005
Ignition Residue 0.001
CAS Max % 1
UN Number UN2811
Percent Purity 98.5 to 101.5%