Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketotifen Hydrogen Fumarate, EP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
34580-14-8
C23H23NO5S
425.50
YNQQEYBLVYAWNX-WLHGVMLRSA-N
(2E)-but-2-enedioic acid; 2-(1-methylpiperidin-4-ylidene)-6-thiatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3(7),4,10,12-pentaen-8-one
OC(=O)\C=C\C(O)=O.CN1CCC(CC1)=C1C2=C(SC=C2)C(=O)CC2=CC=CC=C12
34580-14-8
C23H23NO5S
425.50
YNQQEYBLVYAWNX-WLHGVMLRSA-N
(2E)-but-2-enedioic acid; 2-(1-methylpiperidin-4-ylidene)-6-thiatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3(7),4,10,12-pentaen-8-one
OC(=O)\C=C\C(O)=O.CN1CCC(CC1)=C1C2=C(SC=C2)C(=O)CC2=CC=CC=C12

K3012, 34580-14-8, C19H19NOS•C4H4O4

Spectrum™ Ketotifen Hydrogen Fumarate, EP is used to treat allergic conjunctivitis and to prevent asthma attacks. The EP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Sulfated Ash 0.001
Loss on Drying 0.005
Quantity 25 g
CAS Max % 1
Percent Purity 98.5 to 101%