Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Aramid Fiber 1800 Medium Weight Gloves

Catalog Number p-7144824
Click to view available options
Size:
Small
Medium
Large
X-Large
Glove Cuff Color:
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Medium weight gloves are constructed of 100% aramid fiber and offer ANSI Level A2 protection.

  • 100% aramid fiber
  • Ambidextrous
  • Made in US
ANSI Abrasion Rating ANSI 1
Certifications/Compliance ASTM F1790-97, ASTM D3884-05
Construction String Knit
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Steel processing, metal handling
Glove Cuff Color Yellow
Model 1800S
Series Medium Weight 1800
Yarn Gauge 7
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit