Learn More
Brady™ Keyed Alike Nylon Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
Supplier: Brady™ 123338
View more versions of this product
Safety Services Available
Overview
LOTO Assessments
Description
- Keyed-Alike padlock with non-conductive, spark resistant lock body
- Compact and lightweight
- Available in a variety of colors
Specifications
Black | |
Nylon | |
Padlock |
Keyed Alike | |
3/Pk. | |
1.5 in. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov