Learn More
Brady™ Keyed-Different Nylon Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
Supplier: Brady™ 123355
View more versions of this product
Safety Services Available
Overview
LOTO Assessments
Description
- Keyed-different padlock with non-conductive, spark resistant lock body
- Compact and lightweight
- Available in a variety of colors
Specifications
Orange | |
Keyed Different | |
Nylon | |
6/Pk. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov