Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Keyed Different Safety Lock with Steel Shackle
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient and cost-effective Safety Lockout Padlocks from Brady.
$10.11 - $246.00
Specifications
Product Type | Padlock |
---|---|
Key Type | Keyed Different |
Material | Nylon |
Diameter (English) Shackle | 0.25 in. |
Diameter (Metric) Shackle | 6.35 mm |
Catalog Number | Mfr. No. | Color | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Quantity | Price | Quantity & Availability | ||||
19-000-685
|
Brady™
SDPLRED38STKD |
Red | 1 Ea. |
Each for $10.21
|
|
||||
19-000-686
|
Brady™
SDPLYLW38STKD |
Yellow | 1 Ea. |
Each for $10.21
|
|
||||
19-000-687
|
Brady™
SDPLBLU38STKD |
Blue | 1 Ea. |
Each for $10.21
|
|
||||
19-000-688
|
Brady™
SDPLORG38STKD |
Orange | 1 Ea. |
Each for $10.21
|
|
||||
19-000-689
|
Brady™
SDPLGRN38STKD |
Green | 1 Ea. |
Each for $10.11
|
|
||||
19-000-690
|
Brady™
SDPLRED38STKD6 |
Red | 6 Pk. |
Pack of 6 for $60.50
|
|
||||
19-000-691
|
Brady™
SDPLYLW38STKD6 |
Yellow | 6 Pk. |
Pack of 6 for $61.50
|
|
||||
19-000-692
|
Brady™
SDPLBLU38STKD6 |
Blue | 6 Pk. |
Pack of 6 for $61.50
|
|
||||
19-000-693
|
Brady™
SDPLORG38STKD6 |
Orange | 6 Pk. |
Pack of 6 for $60.50
|
|
||||
19-000-694
|
Brady™
SDPLGRN38STKD6 |
Green | 6 Pk. |
Pack of 6 for $60.50
|
|
||||
19-000-700
|
Brady™
SDPLRED38STKD12 |
Red | 12 Pk. |
Pack of 12 for $123.50
|
|
||||
19-000-702
|
Brady™
SDPLRED38STKD24 |
Red | 24 Pk. |
Pack of 24 for $246.00
|
|
||||
Description
- Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program.
- Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
- Nylon padlocks with steel shackles are lightweight, durable and ideal for general lockout applications.
- Locks that are Keyed Different (KD) are ideal for simple lockout applications and a manageable number of energy isolation points.
- Record and retain engraved key codes to streamline replacement of lost keys.
Specifications
Padlock | |
Nylon | |
6.35 mm | |
Ideal for electrical contractors, maintenance, electricians | |
Steel |
Keyed Different | |
0.25 in. | |
0.82 x 1.5 x 1.77 in. | |
38.10 mm | |
6 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title