Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Heavy Duty Polypropylene Cleanroom Lab Coats
SDP

Catalog No. 19068434
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight, latex-free lab coats are made from high quality materials and compatible in Class ISO7 cleanroom environments.

Protect workers against dry particulates, dirt and grime

  • Non-woven, nonlinting, spunbond heavyweight polypropylene construction
  • Serged seams offer high tensile strength
  • Latex-free
  • Disposable
  • Elastic wrists, snap front, single collar, no pockets
  • Available in six sizes
  • Flame resistant

Ideal for Class 10,000 environments and above. Commonly used in electronic, medical device and pharmaceutical industries where operator garment control is a concern.

Specifications

Product Type Controlled Environments Lab Coat
Color White
Closure Type Snaps
Disposable Single-use
Collar Type Single Ply
Seam Type Serged
For Use With (Application) Electronics, Medical Device, Pharmaceutical
Cleanroom Class Class 10,000 (ISO 7)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.