Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone™ Laminated Polypropylene Water-Repellant Shoe Cover with Non-Skid AQ Sole

Manufacturer:  KEYSTONE ADJUSTABLE CAP COMPANY INC SCNWPIAQCR2XL

 View more versions of this product

Catalog No. 19-550-013


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Polypropylene
Shoe Covers
13cm
15.25 x 5.12 in. (38.73 x 13cm)
Non-skid AQ
Manufacturing, food service, industrial, medical, clean room, pharmaceutical
Yes
Non-skid AQ sole
5.12 in.
White
2XL
Polypropylene
15.25 in.
38.73cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit