Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone™ Polypropylene Shoe Covers - Heavy Duty, Non-Skid

$195.88 - $221.93

Specifications

Coating Vinyl
Material Polyethylene
Resistance Fluid
Sole Type Non-skid
View More Specs

Products 2
Catalog Number Mfr. No. Color Size Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Weight (English) Weight (Metric) Price Quantity & Availability  
19-055-182
View Documents
Keystone™
SCCPEHDXL
White, Blue X-Large 9 lb. 4.08kg
Case of 150 for $221.93
Add to cart
 
19-055-183
View Documents
Keystone™
SCCPEHDLGB
Blue Large 8 lb. 3.62kg
Case of 300 for $195.88
Add to cart
 
Specifications

Specifications

Vinyl
Polyethylene
Fluid
Non-skid
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit