Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Polypropylene Shoe Covers
SDP

Catalog Number 19055176
Click to view available options
Size:
Large
Universal
Color:
Blue
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for contamination control and safety

 

  • 100 pair/case
  • White or Blue 

 

Size Large
Material Polypropylene
Color White
For Use With (Application) Ideal for contamination control and safety
Packaging Quantity 100 Pairs/Cs.
Resistance Fluid
Product Type Controlled Environments Shoe Covers
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit