Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Keystone Safety Super Sticky™ Polypropylene Shoe Cover
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
Description
- Available in different sizes and colors
- Made up of polypropylene
Specifications
Size | Medium |
Material | Polypropylene |
Coating | EVA-PE Film |
Color | White |
Sole Type | Non-skid |
Construction | Polypropylene |
For Use With (Application) | Manufacturing, food service, industrial, medical, clean room, pharmaceutical |
Product Type | Controlled Environments Shoe Covers |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction