Keystone™ Super Sticky™ Shoe Cover

Manufacturer: KEYSTONE ADJUSTABLE CAP COMPANY INC SCSSMEDBL

 View more versions of this product

Catalog No. 19-061-441

  Check Availability
Add to Cart

    Specifications

    Size Medium
    Sole Type Non-skid
    Product Type Shoe cover
    Material Polypropylene
    For Use With (Application) Manufacturing, food service, industrial, medical, clean room, pharmaceutical
    Latex Free Yes
    Coating EVA-PE Film
    Color Blue
    Construction Spunbound Polypropylene with EVA-PE film
    Includes Coated EVA-PE film

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit