Learn More
Keystone™ Tyvek Covers for Containers
Steam component bags
Manufacturer:  Keystone™ COVTVKHS20X40
 View more versions of this product
Catalog No. 19-061-309
Add to cart
Description
Description
  • High density polyethylene
  • Resistant to tears and punctures
  • Flexible and smooth
  • Excellent strength
  • Low-linting
  • Chemical resistant
  • Water resistant
  • Lightweight
  • Opaque
Specifications
Specifications
51 x 102cm (20 x 40 in.)
Cover
HDPE
Containers
Opaque
Safety