Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cumene, 99%, pure
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -96.0°C |
| Color | Colorless |
| Density | 0.8620g/mL |
| Boiling Point | 152.0°C to 154.0°C |
| Flash Point | 31°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4905 to 1.4925 |
| Linear Formula | C6H5CH(CH3)2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Flammable liquid and vapour. May be fatal if swallowed and enters airways. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do NOT induce vomiting. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and keep at rest in a position comfortabl |
| EINECSNumber | 202-704-5 |
| RTECSNumber | GR8575000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction