Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sudan Black, Fisher BioReagents™
Shop All Fisherbrand Products
Click to view available options
Quantity:
10 g
Description
Sudan Black is used to stain chromosomes and lipoproteins, as well as fat in bacteria.
Specifications
Specifications
| Melting Point | 120°C |
| Extinction Coefficient | 600g(-1) cm(-1) min. |
| Merck Index | 15, 9016 |
| Identification | Pass Test |
| Lambda Max | 598 to 602nm |
| Formula Weight | 456.54 |
| Loss on Drying | 5% max. |
| Packaging | Glass Bottle |
| Color Index No. | 26150 |
| Color | Brown |
| Show More |
Safety and Handling
| Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction