Learn More
KHDRBS2 Mouse anti-Human, Mouse, DyLight 650, Clone: 7G8C10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237254C
Catalog No. NBP237254C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
KHDRBS2
7G8C10
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
KHDRBS2
Purified recombinant fragment of human KHDRBS2 (AA: 160-349) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
hSLM-1, KH domain containing, RNA binding, signal transduction associated 2, KH domain-containing, RNA-binding, signal transduction-associated protein 2, MGC26664, Sam68-like mammalian protein 1, SLM1bA535F17.1, SLM-1FLJ38664
Mouse
IgG1
0.1 ml
Primary
202559