Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KHDRBS3 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB29889
Description
Rabbit polyclonal antibody raised against partial synthetic protein of human KHDRBS3.
- High specificity
- High reactivity
Specifications
KHDRBS3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial synthetic protein of human KHDRBS3. | |
In 1X PBS , pH 7.4 (2% sucrose, 0.09% sodium azide). | |
Etle/SALP/SLM-2/SLM2/T-STAR/TSTAR/etoile | |
Rabbit | |
Affinity purification | |
RUO | |
10656 | |
Store at 4°C for up to one week. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:250) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
O75525 | |
KHDRBS3 | |
A synthetic peptide corresponding to amino acids 281-330 of human KHDRBS3. | |
100 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction