Tocris™ Ki 16425

LPA1 and LPA3 receptor antagonist

Manufacturer: Tocris™ 5056/50

 View more versions of this product

Catalog No. 505650R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors Ki 16425
    Purity >98
    Molecular Weight 474.96
    Quantity 50mg
    Product Type Ki 16425

    Antagonist of the lysophosphatidic acid receptors LPA1 and LPA3 (Ki values are 0.25 and 0.36µM respectively, in a GTPγS binding assay). Blocks LPA-induced dephosphorylation of Yes-associated protein (YAP) and WW domain-containing transcription regulator protein 1 (TAZ), inhibiting the Hippo signaling pathway, in HEK293A cells.

    • Chemical Name: 3-[[[4-[4-[[[1-(2-Chlorophenyl)ethoxy]carbonyl]amino]-3-methyl-5-isoxazolyl]phenyl]methyl]thio]propanoic acid
    • Formula: C23H23ClN2O5S
    • CAS Number: 355025-24-0
    • Solubility: Soluble to 100mM in DMSO and to 20mM in 1eq. NaOH
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit