Learn More
Selleck Chemical LLCSupplier Diversity Partner Ki8751 10mg 228559-41-9  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S136310MG
Ki8751 is a potent and selective inhibitor of VEGFR2 with IC50 of 0.9 nM, >40-fold selective for VEGFR2 than c-Kit, PDGFRα and FGFR-2, little activity to EGFR, HGFR and InsR.
Catalog No. 50-712-8