Learn More
Abnova™ KIAA0101(variant 1) 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00009768T01
Catalog No. 89-949-644
Add to cart
Specifications
Specifications
293T
NM_014736.4
pCMV-KIAA0101(variant1) full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KIAA0101(variant1) 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 12.32kDa
Transient overexpression cell lysate was tested with anti-KIAA0101 (variant 1) antibody (Mfr. No. H00009768-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human