Learn More
Abnova™ KIAA0141 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00009812T01
Catalog No. 89-949-648
Add to cart
Description
Description
Specifications
Specifications
293T
NM_014773
pCMV-KIAA0141 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KIAA0141 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 55.9kDa
Transient overexpression cell lysate was tested with anti-KIAA0141 antibody (Mfr. No. H00009812-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human