Learn More
KIAA0182 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 249471AP
Catalog No. 50-173-4636
Add to cart
Description
Description
This gene encodes a proline-rich protein with coiled coil domains that may be a subunit of a BRAF35-HDAC (BHC) histone deacetylase complex. This gene may function as an oncogene in breast cancer and enhanced expression of the encoded protein has been observed in breast cancer patients.
Specifications
Specifications
KIAA0182
0.44 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14687, Q3U3C9
Genetic suppressor element 1, GSE1, KIAA0182
Rabbit
IgG
150 μL
-20°C
Primary
23199, 382034
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GSE1
Liquid
Gse1
KIAA0182 Fusion Protein Ag20674
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse