Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0319 Mouse anti-Human, Clone: PCRP-CREB5-1G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30747710UG

 View more versions of this product

Catalog No. NB122427


Add to cart

Description

Description

KIAA0319 Monoclonal antibody specifically detects KIAA0319 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

KIAA0319
PCRP-CREB5-1G8
Unconjugated
10 mM PBS with 0.05% BSA
DLX2, DYLX2, dyslexia susceptibility 2, DYX2, KIAA0319, MGC176717
Recombinant full-length human KIAA0319 protein
Protein A or G purified
RUO
Primary
9856
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
0.2 mg/ml
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Purified
Mouse
IgG2b Kappa
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit