Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0319 Mouse anti-Human, Clone: PCRP-CREB5-1G8, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen KIAA0319
Clone PCRP-CREB5-1G8
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122428
View Documents
Novus Biologicals
NBP30747720UG
20 μg
Each for $138.37
Add to cart
 
NB122427
View Documents
Novus Biologicals
NBP30747710UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

KIAA0319 Monoclonal antibody specifically detects KIAA0319 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

KIAA0319
0.2 mg/ml
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
9856
IgG2b Kappa
Primary
Store at 4°C.
PCRP-CREB5-1G8
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Human
DLX2, DYLX2, dyslexia susceptibility 2, DYX2, KIAA0319, MGC176717
Recombinant full-length human KIAA0319 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit