Learn More
Aviva Systems Biology Corp KIAA0494 Antibody - N-terminal region (ARP47075_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47075_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0494. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA0494. Alternate names: RP11-8J9.3, KIAA0494ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59467