Learn More
KIAA0494 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB30042
Catalog No. 89-033-330
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human KIAA0494.
  • High specificity
  • High reactivity
Specifications
Specifications
KIAA0494
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user.
NM_014774
RP11-8J9.3
Rabbit
Protein A purification
RUO
Primary
9813
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human KIAA0494.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
KIAA0494
A synthetic peptide corresponding to N-terminus of human KIAA0494.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)