Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0495 Mouse anti-Human, Clone: OTI2A9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503634

Catalog No. 50-167-4480


Add to cart

Description

Description

TP73-AS1 in molecular biology is P73 antisense RNA 1T (non-protein coding) and also known as PDAM, is a long non-coding RNA. It may regulate apoptosis via regulation of p53-dependent anti-apoptotic genes. It may play a role in the development of oligodendroglial tumours.
Specifications

Specifications

KIAA0495
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UF72
KIAA0495, PDAM
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57212
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2A9
Unconjugated
TP73-AS1
Liquid
TP73-AS1
Full length human recombit protein of human KIAA0495 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit