Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0513 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP304655100UL

 View more versions of this product

Catalog No. NB095067


Add to cart

Description

Description

KIAA0513 Polyclonal antibody specifically detects KIAA0513 in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

KIAA0513
Unconjugated
Purified
Rabbit
IgG
100 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Mouse, Rat
Western Blot
PBS with 50% glycerol, pH7.3.
hypothetical protein LOC9764, KIAA0513
Recombinant fusion protein containing a sequence corresponding to amino acids 1-301 of human KIAA0513 (NP_001273494.1).
Affinity purified
RUO
Primary
9764
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit