Learn More
Aviva Systems Biology Corp KIAA0515 Antibody - N-terminal region (ARP47973_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47973_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0515. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIAA0515. Alternate names: RP11-334J6.1, DKFZp781F05101, DKFZp781K12107, LQFBS-1, MGC10526, BAT2L, BAT2L1, KIAA0515ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59690