Learn More
Aviva Systems Biology Corp KIAA0892 Antibody - middle region (ARP55232_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55232_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA0892. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA0892. Alternate names: MAU2L, MGC75361, mau-2, SCC4, KIAA0892ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61442